missing translation for 'onlineSavingsMsg'
Learn More

Dess-Martin Periodinane (Technical Grade), TRC

Product Code. 30011441
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011441 25 g 25g
30007579 100 g 100g
30010473 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011441 Supplier Toronto Research Chemicals Supplier No. TRCD29742025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87413-09-0
Molecular Formula C13H13IO8
Molecular Weight (g/mol) 424.14
Synonym Acetic Acid 1,1',1' '-(3-Oxo-1lambda5-1,2-benziodoxol-1(3H)-ylidyne) Ester, 1,1,1-Tris(acetyloxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Dess-Martin Reagent, Martin's Reagent, Triacetoxyperiodinane, DMP
IUPAC Name (1,1-diacetyloxy-3-oxo-1λ^{5},2-benziodoxol-1-yl) acetate
SMILES CC(=O)OI1(OC(=O)C)(OC(=O)C)OC(=O)c2ccccc12

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H13IO8/c1-8(15)19-14(20-9(2)16,21-10(3)17)12-7-5-4-6-11(12)13(18)22-14/h4-7H,1-3H3
Formula Weight 423.9655
Chemical Name or Material Dess-Martin Periodinane (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.