missing translation for 'onlineSavingsMsg'
Learn More

Dess-Martin periodinane

Product Code. 11362777
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362777 1 g 1g
11372777 5 g 5g
11439840 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362777 Supplier Thermo Scientific Alfa Aesar Supplier No. L15779.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87413-09-0 | C13H13IO8 | 424.14 g/mol

Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.

Solubility
Soluble in chloroform, acetone, acetonitrile and methylene chloride. Slightly soluble in ether and hexane.

Notes
Store in cool place. Air and moisture sensitive. Incompatible with strong reducing agents, combustible material and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87413-09-0
Molecular Formula C13H13IO8
Molecular Weight (g/mol) 424.14
MDL Number MFCD00130127
InChI Key NKLCNNUWBJBICK-UHFFFAOYSA-N
PubChem CID 159087
SMILES CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12

Specifications

Melting Point 130°C to 132°C (decomposition)
Quantity 1 g
UN Number UN1479
Beilstein 4548207
Sensitivity Air, light, and moisture sensitive
Merck Index 14,2933
Synonym dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
Solubility Information Soluble in chloroform,acetone,acetonitrile and methylene chloride. Slightly soluble in ether and hexane.
IUPAC Name (1,1-diacetyloxy-3-oxo-1$l^{5},2-benziodoxol-1-yl) acetate
Formula Weight 424.15
Percent Purity ≥95%
Chemical Name or Material Dess-Martin periodinane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.