Learn More
Brady™ Destructible Tamper-evident Matte Vinyl Labels
Identify product tampering and prevent one-piece label removal with the B-351 tamper-resistant vinyl label material. Easily print almost anywhere with a Brady™ portable label printer.
Brand:  Brady™ 170856
 View more versions of this product
Product Code. 17871749
  • 65.25 € / Pack of 360
In Stock
Add to basket
Description
Description
  • For use in asset tracking, rating plates, product security and tamper identification
  • Designed to fracture to show signs of product tampering and prevent one-piece removal
  • Made with a permanent adhesive that bonds tightly to almost any surface
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
White
Die-cut
Top
Label
Asset tracking;Security identification
1
M5-17-351
360
-40°F to 176°F
0.0028 in
25.40 mm (W) x 12.70 mm (H)
1 in.
25.40 mm
0.5 in.
M5-17-351
2011/65/EU, 2015/863
None
Matte
Vinyl
Thermal Transfer
Non-sterile
BMP51;BMP53;M511;M510
-40°C
360 Label(s) / Box
25.40 mm (W) x 12.70 mm (H)
-40°C to 80°C
0.071 mm
0.5 in.
25.40 mm
0.5 in
12.70 mm