missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Detergent Removal Spin Columns & Plates

Product Code. 10792447 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
2 x 96-well Plates
24 x 0.1 mL Columns
25 x 125 μL Columns
25 x 500 μL Columns
5 mL
5 x 2 mL Columns
5 x 4 mL Columns
Product Line:
HiPPR
Pierce
Product Type:
Detergent Removal Resin
Spin Column
Spin Plate
Unit Size:
1 set
10mL
Pack of 2
Pack of 24
Pack of 25
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Line Product Type Unit Size
10792447 5 x 2 mL Columns Pierce Spin Column Pack of 5
10116434 10 mL Pierce Detergent Removal Resin 10mL
10645585 25 x 125 μL Columns Pierce Spin Column Pack of 25
10499764 25 x 500 μL Columns Pierce Spin Column Pack of 25
11736446 5 x 4 mL Columns Pierce Spin Column Pack of 5
11368261 2 x 96-well Plates Pierce Spin Plate Pack of 2
12301883 5 mL HiPPR Spin Column 1 set
12391873 24 x 0.1 mL Columns HiPPR Spin Column Pack of 24
12201233 2 x 96-well Plates HiPPR Spin Plate Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10792447 Supplier Thermo Scientific™ Supplier No. 87778
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly eliminate high concentrations of detergents with minimal sample loss with detergent removal resins. Thermo Scientific HiPPR (High Protein and Peptide Recovery) resin is recommended for protein concentrations up to 100 μg/mL, while Thermo Scientific Pierce resin is suitable for concentrations over 100 μg/mL.

Detergents and surfactants used to prepare protein and peptide samples can interfere with analysis by ELISA, isoelectric focusing, and mass spectrometry (MS). Removing detergents from peptide samples is especially challenging and critical for MS analysis. Even low concentrations of detergents will contaminate instruments and interfere with column binding, elution, and peptide ionization. Thermo Scientific HiPPR and Pierce detergent removal resins are ideal for rapid removal of common detergents from tryptic digests to improve the results of LC-MS/MS and MALDI-MS analysis.

  • Optimized—removes >95% of detergent from samples of proteins or peptides
  • Fast—sample processing takes less than 15 minutes
  • Effective—eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing, and mass spectrometry

Pierce Detergent Removal Resin

Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 μL to 1 mL samples with minimal sample loss. These microcentrifuge columns come prepacked with various volumes of proprietary resin and are best for processing samples with proteins or peptide concentrations greater than 100 μg/mL.

HiPPR Detergent Removal Spin Column Kit

HiPPR Detergent Removal resin has been optimized for sample concentrations of 1 to 100 μg/mL and is ideal for removing commonly used detergents, including SDS, Triton™ X-100, NP-40 and CHAPS at concentrations of 0.5-1%. The resin is available in pre-filled spin columns, 96-well filter spin plates, and as loose resin for customization.

Available formats:

  • Resin slurries—bulk resin supplied with empty 0.8-mL spin columns
  • Spin columns—0.1 mL micro-centrifuge columns
  • 96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification
TRUSTED_SUSTAINABILITY

Specifications

Description 15 mL Conical, Spin Column
Product Type Spin Column
Content And Storage Upon receipt store at 4°C. The resin is stored in 0.15 M NaCl, 0.05% sodium azide.
Column Type Proprietary Resin, Affinity
Stationary Phase Proprietary Ligand
Product Line Pierce
Purification Target Detergent Removal
Sufficient For 150 to 500 μL Sample Each Column
Quantity 5 x 2 mL Columns
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.