missing translation for 'onlineSavingsMsg'
Learn More

Deuterium chloride, 20% w/w in D{2}O, 99.5% (Isotopic)

Product Code. 15476455
Change view
Click to view available options
Quantity:
50 g
Packaging:
Clear Glass Ampule
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15476455 50 g Clear Glass Ampule 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15476455 Supplier Thermo Scientific Alfa Aesar Supplier No. 042407.18
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7698-05-7 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 1.2 g/cm3 at 20°C
Odor Acidic
Packaging Clear Glass Ampule
Refractive Index 1.373
Sensitivity Moisture
Quantity 50 g
CAS 7698-05-7
Boiling Point 108°C
Assay Percent Range 99.5% (Isotopic)
Molecular Formula ClH
MDL Number MFCD00011266
Synonym Hydrogen chloride, deuterated
Solubility Information Fully miscible in water.
InChI Key VEXZGXHMUGYJMC-FTGQXOHASA-N
SMILES [1H+].[Cl-]
IUPAC Name proton chloride
Molecular Weight (g/mol) 36.46
Formula Weight 37.47
Chemical Name or Material Deuterium chloride, 20% w/w in D2O
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.