missing translation for 'onlineSavingsMsg'
Learn More

Deuterium chloride, for NMR, 1M solution in D2O, 99.8 atom% D, AcroSeal™

Product Code. 10566864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10566864 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10566864 Supplier Thermo Scientific Acros Supplier No. 210601000
  • ONLINE EXCLUSIVE PRICE
    242.00 €
    234.00 € / 100mL
    Save 8.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7698-05-7 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Density 1.1400g/mL
Deuterium (D) 99.8% min. (NMR)
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
CAS 7698-05-7
Molecular Formula ClH
Linear Formula DCl
MDL Number MFCD00044642
Synonym deuterium chloride, hydrochloric acid-d, deuterium chloride, 20 wt% in deuterium oxide, chlorohydrogen, hydrochloric acid d, chloro 2h, deuterium chloride, 99 atom % d, deuterium chloride, 1 m in deuterium oxide, deuterium chloride, 0.1 m in deuterium oxide, deuterium chloride, 38 wt% in deuterium oxide
InChI Key VEXZGXHMUGYJMC-FTGQXOHASA-N
SMILES Cl
Molecular Weight (g/mol) 36.46
PubChem CID 522634
Formula Weight 37.47
Grade NMR
Chemical Name or Material Deuterium chloride, 1M solution in D2O, 99.8 atom% D
Show More Show Less
compliance-icons
Product Identifier
  • Deuterium chloride, 1M solution in D2O
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Deuterium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.