missing translation for 'onlineSavingsMsg'
Learn More

Deuterium oxide, for NMR, 100.0 atom % D

Product Code. 10760671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
Packaging:
Glass Bottle
Septum Bottle
Unit Size:
100mL
10mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10760671 10 mL Glass Bottle 10mL
16338186 10 mL Septum Bottle 10mL
10265880 50 mL Glass Bottle 50mL
10275880 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10760671 Supplier Thermo Scientific Acros Supplier No. 166310100
  • ONLINE EXCLUSIVE PRICE
    74.00 €
    71.80 € / 10mL
    Save 2.20 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7789-20-0 | H2O | 20.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-20-0
Molecular Formula H2O
Molecular Weight (g/mol) 20.03
MDL Number MFCD00044636
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
Synonym deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
PubChem CID 24602
ChEBI CHEBI:41981
IUPAC Name [(²H)oxy](²H)
SMILES [2H]O[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 1.1050g/mL
Deuterium (D) 99.95% min.
Packaging Glass Bottle
Refractive Index 1.328
Specific Gravity 1.105
Viscosity 1.25 mPa.s (20°C)
Quantity 10 mL
Melting Point 3.8°C
Boiling Point 101.4°C
Linear Formula D2O
Merck Index 15, 2942
Solubility Information Solubility in water: miscible, exothermic reaction.
Formula Weight 20.02
Grade NMR
Chemical Name or Material Deuterium oxide, 100.0 atom % D
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.