missing translation for 'onlineSavingsMsg'
Learn More

Deuterium oxide, for NMR, packaged in 0.75ml ampoules, 99.8 atom% D

Product Code. 10254651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10254651 7.5 mL 7.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10254651 Supplier Thermo Scientific Acros Supplier No. 351430075
  • ONLINE EXCLUSIVE PRICE
    33.40 €
    30.70 € / 7.5mL
    Save 2.70 €
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7789-20-0 | H2O | 20.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 1.1050g/mL
Deuterium (D) 99.75% min.
Refractive Index 1.328
Specific Gravity 1.105
Viscosity 1.25 mPa.s (20°C)
Quantity 7.5 mL
CAS 7789-20-0
Melting Point 3.8°C
Boiling Point 101.4°C
Molecular Formula H2O
Linear Formula D2O
MDL Number MFCD00044636
Merck Index 15, 2942
Synonym deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
Solubility Information Solubility in water: miscible, exothermic reaction.
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES [2H]O[2H]
IUPAC Name [(²H)oxy](²H)
Molecular Weight (g/mol) 20.03
PubChem CID 24602
ChEBI CHEBI:41981
Formula Weight 20.02
Grade NMR
Chemical Name or Material Deuterium oxide, 99.8 atom% D
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.