missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Devarda's Alloy, granular

Product Code. 15472337
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472337 500 g 500g
15462337 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15472337 Supplier Thermo Scientific Chemicals Supplier No. 045526.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 8049-11-4 | Al9Cu10Zn | 943.67 g/mol

Devarda's Alloy is used as reducing agent in analytical chemistry for the determination of nitrates after their reduction to ammonia under alkaline conditions. It is also used in the quantitative or qualitative analysis of nitrates in agriculture and soil science before the development of ion chromatography. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8049-11-4
Molecular Formula Al9Cu10Zn
Molecular Weight (g/mol) 943.67
MDL Number MFCD00198176
InChI Key MPKKOQSNGWAMJD-UHFFFAOYSA-N
Synonym devarda's alloy, devarda's alloy, granular, al.cu.zn, devarda's alloy, p.a., alumane copper zinc, devarda's alloy, puriss. p.a., devarda's alloy, saj special grade, devarda's alloy, powder,-100 mesh, devarda's alloy, filings, +100 mesh, devarda inverted exclamation mark s alloy
PubChem CID 16211762
IUPAC Name aluminum;copper;zinc
SMILES [Al].[Cu].[Zn]

Specifications

Density 4.355
Quantity 500 g
Form Granular
Solubility Information Insoluble in water. Slightly soluble in hydrochloric acid
Chemical Name or Material Devarda's Alloy
compliance-icons
Product Identifier
  • Devarda's Alloy, granular
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Devardas alloy, granular

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.