missing translation for 'onlineSavingsMsg'
Learn More

Devarda's Alloy, Granular, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15548044 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15548044 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548044 Supplier Avantor J.T.Baker Supplier No. 268001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 8049-11-4
Quantity 500 g
Packaging Poly Bottle
Form Granular
Molecular Formula Al9Cu10Zn
Synonym devarda's alloy, devarda's alloy, granular, al.cu.zn, devarda's alloy, p.a., alumane copper zinc, devarda's alloy, puriss. p.a., devarda's alloy, saj special grade, devarda's alloy, powder,-100 mesh, devarda's alloy, filings, +100 mesh, devarda inverted exclamation mark s alloy
InChI Key MPKKOQSNGWAMJD-UHFFFAOYSA-N
SMILES [Al].[Cu].[Zn]
IUPAC Name aluminum;copper;zinc
Molecular Weight (g/mol) 943.67
PubChem CID 16211762
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Devarda's Alloy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.