missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Devazepide

Product Code. 18025162
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18025162 10 mg 10mg
18014092 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025162 Supplier R&D Systems™ Supplier No. 2304/10
  • ONLINE EXCLUSIVE PRICE
    406.17 €
    362.25 € / 10mg
    Save 43.92 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective, orally active CCK1 receptor antagonist

Potent, orally active CCK1 (CCK-A) receptor antagonist that displays appetite-stimulant effects. Blocks the anorectic response to CCK-8 and increases food intake in rats following systemic and i.c.v administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103420-77-5
Molecular Formula C25H20N4O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 408.46
Purity >99%
Molecular Weight (g/mol) 408.461
Chemical Name or Material Devazepide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.