missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dexamethasone, 10 mM in DMSO

Product Code. 12063755 Shop All Gibco Products
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12063755 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12063755 Supplier Gibco™ Supplier No. A13449
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This solution contains 10 mM dexamethasone in DMSO. Dexamethasone is an anti-inflammatory glucocorticoid used as a supplement in hepatocyte cell culture.

  • Solution contains 10 mM dexamethasone in DMSO
  • Dexamethasone is an anti-inflammatory glucocorticoid used as a supplement in hepatocyte cell culture
  • This product is available as a separate component for the purposes of supplementing William's E Medium
  • No phenol red (A1217601) for the maintenance of Human Spheroid-Qualified Hepatocytes in 3D
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2 to 8°C) and protect from light
Cell Type Hepatocyte Cells
Concentration 10 mM
Form Liquid
Product Type DMSO
Without Additives No Phenol Red
For Use With (Application) Cell Culture
Quantity 50 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.