missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone 9,11-Epoxide, TRC

Product Code. 30011519
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011519 1 g 1g
30009965 100 mg 100mg
30006167 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011519 Supplier Toronto Research Chemicals Supplier No. TRCD2987951G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24916-90-3
Molecular Formula C22 H28 O5
Molecular Weight (g/mol) 372.45
Synonym 17,21-Dihydroxy-16α-methyl-9β,11β-epoxypregna-1,4-diene-3,20-dione, 9β,11β-Epoxy-17,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, Dexamethasone 9,11-Epoxide
IUPAC Name 17,21-Dihydroxy-16α-methyl-9β,11β-epoxypregna-1,4-diene-3,20-dione
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H28O5/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15)18(27-22)10-20(16,3)21(12,26)17(25)11-23/h6-7,9,12,15-16,18,23,26H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,18+,19+,20+,21+,22-/m1/s1
Formula Weight 372.1937
Percent Purity >95%
Chemical Name or Material 9beta,11beta-Epoxy-17,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.