missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar A17590.06

Product Code. 11418217

  • 433.00 € / 5g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Anti-inflammatory glucocorticoidDexamethasone is used to treat inflammatory and autoimmune conditions such as rheumatoid arthritis and bronchospasm. It is useful to study apoptosis, cell signaling pathways and gene expression. It is associated with marbofloxacin and clotrimazole and finds application in veterinary medicine to treat difficult ear infections in dogs. It is also used to treat horses with swelling of of distal limbs and general bruising in combination with trichlormethiazide. It is also an anti-inflammatory glucocorticoid.

Solubility
Soluble in ethanol and dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool place. Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50-02-2
C22H29FO5
5g
Light sensitive
dexamethasone, decadron, dexamethazone, maxidex, decaspray, desametasone, hexadecadrol, dexacort, dexasone, hexadrol
UREBDLICKHMUKA-CXSFZGCWSA-N
(1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
5743
392.47
Dexamethasone
0.98
MFCD00064136
2066651
142943
Soluble in ethanol and dimethyl sulfoxide. Insoluble in water.
C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
392.47
CHEBI:41879
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.