missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexamethasone, 99.5%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
Specifications
Specifications
| Color | White |
| Assay Percent Range | 0.995 |
| Quantity | 100 mg |
| Formula Weight | 392.47 |
| Physical Form | Powder |
| Chemical Name or Material | Dexamethasone, For HPLC analysis |
Safety and Handling
Product Identifier
- dexamethasone
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : dexamethasone
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction