missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone acetate, MedChemExpress

Product Code. 16461429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461429 1 g 1g
16418727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461429 Supplier Medchem Express Supplier No. HY14648A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dexamethasone acetate (Dexamethasone 21-acetate) is a glucocorticoid receptor agonist. Dexamethasone acetate has the potential for ophthalmic infections treatment.

Dexamethasone acetate (Dexamethasone 21-acetate) is a glucocorticoid receptor agonist. Dexamethasone acetate has the potential for ophthalmic infections treatment.

Specifications

Chemical Name or Material Dexamethasone acetate
CAS 1177-87-3
Color White
Purity Grade Notes Research
Molecular Formula C24H31FO6
Quantity 1 g
Synonym Dexamethasone 21-acetate Hexadecadrol acetate
Solubility Information DMSO : 50 mg/mL (115.07 mM; Need ultrasonic) ∣ H2O : ≥ 1 mg/mL (2.30 mM)
SMILES C[C@@]1([C@@]2(O)C(COC(C)=O)=O)[C@](C[C@H]2C)([H])[C@@](CCC3=CC4=O)([H])[C@@](F)([C@]3(C=C4)C)[C@@H](O)C1
Molecular Weight (g/mol) 434.5
Formula Weight 434.5
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.