missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone, MP Biomedicals™

Product Code. 17118548 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17118548 500 mg 500mg
11474482 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17118548 Supplier MP Biomedicals Supplier No. 0219004090
  • ONLINE EXCLUSIVE PRICE
    223.82 €
    194.25 € / 500mg
    Save 29.57 €
    13% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anti-inflammatory agent

Dexamethasone is a glucocorticoid anti-inflammatory agent. It regulates T cell survival, growth, and differentiation. It also inhibits the induction of nitric oxide synthase.

Specifications

Chemical Name or Material Dexamethasone
CAS 50-02-2
Synonym maxidex
Quantity 500 mg
Molecular Formula C22H29FO5
Formula Weight 392.47
Physical Form Powder
Color White
compliance-icons
Product Identifier
  • Dexamethasone
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: dexamethasone

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.