missing translation for 'onlineSavingsMsg'
Learn More

Dextran, MP Biomedicals™

Product Code. 11407860 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
Unit Size:
100g
10g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407860 10 g 10g
11417860 50 g 50g
11427860 100 g 100g
11437860 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11407860 Supplier MP Biomedicals Supplier No. 0210150810
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White
Physical Form Powder
Quantity 10 g
Formula Weight 35000 to 50000
Chemical Name or Material Dextran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.