Learn More
Dextran, MW ca 20,000
Description
Low molecular weight dextran is useful as a stabilizer, volume extender, lubricant, binding platform, and matrix component. Higher molecular weight dextrans have differing viscosities, aggregation and permeation properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Merck Index | 14,2948 |
| Solubility Information | Soluble in water (100mg/ml),DMSO (freely),formamide (freely),ethylene glycol (freely),and glycerol (freely). |
| Formula Weight | ≈20 kd |
| Chemical Name or Material | Dextran |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.