Learn More
Dextran, MW ca 6,000, Thermo Scientific™
Dextran, also known as manninotriose, CAS # 9004-54-0, is a glucose polymer of bacterial origin.
Brand:  Thermo Scientific Alfa Aesar J62775.22
197.70 EUR valid until 2026-09-25
BEST PRICE on Promo! Use promo code "27966" to get your promotional price.
 View more versions of this product
Product Code. 15415079
  • 200.00 € / 100g
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Dextran is a complex branched polysaccharide of bacterial origin
• It primarily contains alpha-D C-1 – C-6 glycosidic linkages
• Alternate linkages account for the complex branching of dextran
Application
• Some research suggests that dextran can be used a carrier that helps target delivery of compounds to specific cells
• It is intended for protein isolation and centrifugation protocols, as well as other research applications
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Promotions
Promotions
Promo
Expires: 2026-09-25
Curiosity Goes Beyond
Promo Code: 27966
Push the boundaries of discovery with exceptional savings across essential research workflows: Cell Culture and Analysis driven by our Gibco™ product portfolio, Gene Expression powered by Applied Biosystems™ solutions, and Protein Expression enabled by Invitrogen™ and Thermo Scientific™ products.
Applying Promo and Adding...