missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate (6000 to 8000), MP Biomedicals™

Product Code. 11417880 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417880 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11417880 Supplier MP Biomedicals Supplier No. 0210151850
  • ONLINE EXCLUSIVE PRICE
    598.00 €
    567.00 € / 50g
    Save 31.00 €
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as an anticoagulant, antiviral and antilipemic agent

Dextran sulfate is a polyanionic derivative of dextran produced by esterification of Dextran with chlorosulphonic acid. Dextran sulfate sodium (DSS) is the sodium salt which is a white to off-white powder freely soluble in water and salt solutions to form a stable, clear solution.

  • Sulfur content is approximately 17% which corresponds to an average of 1.9 sulfate groups per glucosyl residue of the dextran molecule
  • High purity and consistent quality enables its application in pharmaceuticals and biotechnology industry

Specifications

Chemical Name or Material Dextran sulfate
Color White to Off-white
pH 5.0 to 7.5 (1% aqueous solution)
CAS 9011-18-1
Physical Form Powder
Quantity 50 g
Molecular Formula (C6H7O5)m(C6H7O5)n
MDL Number MFCD00081551
Synonym dextran sulfate sodium, DSS
Solubility Information Soluble in water
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
IUPAC Name Dextran sulfate sodium salt
Molecular Weight (g/mol) 40,000
Formula Weight ≈1,400,000
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.