missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt, Colitis Grade (36,000 to 50,000), MP Biomedicals™

Product Code. 11424352 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424352 10 g 10g
15722262 25 g 25g
11434352 50 g 50g
12871781 100 g 100g
11454352 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11424352 Supplier MP Biomedicals Supplier No. 0216011010
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Induces severe colitis (inflammatory bowel disease) in laboratory animals

Dextran sulfate sodium salt, or DSS, is a synthetic sulfated branched polysaccharide derivative of dextran that has multiple uses in biomedical and clinical research. One important property of DSS is to trigger colitis in mice and rats when administered in drinking water. DSS binds to medium-chain-length fatty acids in the colon and induces intestinal inflammation. Colitis-grade DSS (MW = 36,000 to 50,000) from MP Bio is one of the most widely used products based on peer reviewed scientific publications.

  • Induces severe colitis (inflammatory bowel disease)
  • Highest sulfur content – 19%
  • Highest chirality - +104° of specific rotation
  • Lowest pH – 6.2 at 1% solution
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dextran Sulfate Sodium Salt
Color White
pH 5 to 7.11 (1% aqueous solution)
CAS 9011-18-1
Physical Form Powder
Boiling Point 310°C
Melting Point 92°C
Molecular Formula (C6H7O5)m(C6H7O5)n
MDL Number MFCD00081551
Solubility Information Soluble in water
Quantity 10 g
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
IUPAC Name Dextran sulfate sodium salt
Molecular Weight (g/mol) 40,000
For Use With (Application) Used for inducing ulcerative colitis in animals and extensively to study colon cancer developing in relation to colonic inflammation
Grade Colitis Grade
Moisture <10%
Packaging Plastic Container
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.