missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt (36,000-50,000 M.Wt), MP Grade, MP Biomedicals™

Product Code. 11414352 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414352 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414352 Supplier MP Biomedicals Supplier No. 0216011001
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color White
CAS 9011-18-1
Physical Form Powder
Molecular Formula (C6H7O5)m(C6H7O5)n
MDL Number MFCD00081551
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
Quantity 1 g
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
IUPAC Name ethyl 4-aminobenzoate
Molecular Weight (g/mol) 40,000
PubChem CID 2337
ChEBI CHEBI:116735
Formula Weight 36,000 to 50,000
Chemical Name or Material Dextran Sulfate Sodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.