missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt, MW ca >500,000

Product Code. 15445089
Click to view available options
Quantity:
5 g
50 g
100 g
1 kg
Unit Size:
100g
1kg
50g
5g
This item is not returnable. View return policy

Product Code. 15445089

Brand: Thermo Scientific Alfa Aesar J62787.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol

Dextran sulfate sodium is used to study colitis (inflammatory bowel disease) as well as an antiviral agent. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White to Yellow
Physical Form Powder
Merck Index 14,2951
Solubility Information Soluble in water.
Quantity 1 kg
IUPAC Name Dextran sulfate sodium salt
Formula Weight >≈500 kd
Odor Phenol-like
Chemical Name or Material Dextran sulfate sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.