missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt, Reagent grade, MP Biomedicals™

Product Code. 11457870 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457870 100 g 100g
11437870 10 g 10g
11447870 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457870 Supplier MP Biomedicals Supplier No. 0210151680
  • ONLINE EXCLUSIVE PRICE
    1010.00 €
    953.40 € / 100g
    Save 56.60 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 8,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White
Physical Form Powder
Quantity 100 g
IUPAC Name Dextran sulfate sodium salt
Formula Weight 8,000 (Average)
Grade Reagent
Chemical Name or Material Dextran sulfate sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.