missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt

Product Code. 15875118 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
50g
Product Code. Quantity Packaging unitSize
15875118 10 g Plastic bottle 10g
15885118 50 g Plastic bottle 50g
15895118 100 g Plastic bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 15875118

Brand: Thermo Scientific J14489.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Physical Form Powder
Quantity 10 g
Formula Weight 40,000- 50,000
Packaging Plastic bottle
Chemical Name or Material Dextran Sulfate Sodium Salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.