missing translation for 'onlineSavingsMsg'
Learn More

Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents

Product Code. 10764534
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10764534 1 kg Poly Bottle 1kg
10051793 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10764534 Supplier Fisher BioReagents Supplier No. BP3501
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
This item is not returnable. View return policy
Dextrose is often used as a cell culture additive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.1°C
Color White
pH 5.9
Assay Percent Range ≥99.8 %
Packaging Poly Bottle
Linear Formula HOCH2(CHOH)4CHO
Quantity 1 kg
Merck Index 15, 4495
Specific Rotation 52.5 to 53.0° (+ or -)
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 180.08
Vapor Pressure Negligible
Percent Purity ≥99.8%
Grade DNase-, RNase- and Protease-Free
Physical Form Solid
Chemical Name or Material Dextrose Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.