missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Description
Dextrose is often used as a cell culture additive.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 146.1°C |
Color | White |
pH | 5.9 |
Assay Percent Range | ≥99.8 % |
Packaging | Glass Bottle |
Linear Formula | HOCH2(CHOH)4CHO |
Quantity | 500 g |
Merck Index | 15, 4495 |
Specific Rotation | 52.5 to 53.0° (+ or -) |
Solubility Information | 0.005% max. Insoluble Matter |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'