missing translation for 'onlineSavingsMsg'
Learn More

Dextrose, Anhydrous, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15517944 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15517944 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15517944 Supplier Avantor J.T.Baker Supplier No. 192005
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GMP-manufactured dextrose meeting USP specifications.

Specifications

CAS 50-99-7
Packaging Poly Bottle
Molecular Formula C6H12O6
Quantity 2.5 kg
Specific Gravity 1L = 1.54kg
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
SMILES C(C(C(C(C(C=O)O)O)O)O)O
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
Molecular Weight (g/mol) 180.156
PubChem CID 107526
ChEBI CHEBI:42758
Formula Weight 180.16
Percent Purity 97.5 to 102.0%
Grade Multi-Compendial/USP
Physical Form Solid
Chemical Name or Material Dextrose, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.