missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Monohydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Molecular Formula | C18H30N2O14 |
Quantity | 2.5 kg |
Specific Gravity | 1L = 1.54kg |
Synonym | nicotine ditartrate dihydrate, unii-r7m676m8yv, nicotine bitartrate usan, nicotine bitartrate who-dd, nicotine acid tartrate dihydrate, s-3-1-methylpyrrolidin-2-yl pyridine bis 2r,3r-2,3-dihydroxysuccinate dihydrate, 2r,3r-2,3-dihydroxybutanedioic acid; 3-2s-1-methylpyrrolidin-2-yl pyridine; dihydrate, 3-2s-1-methyl-2-pyrrolidinyl pyridine, 2r,3r-2,3-dihydroxybutanedioate 1:2 , dihydrate |
InChI Key | LDMPZNTVIGIREC-HYRPJEHINA-N |
SMILES | O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1 |
IUPAC Name | (2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol hydrate |
Molecular Weight (g/mol) | 498.44 |
Formula Weight | 198.17 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'