Learn More
Applied Biosystems™ dGTP BigDye™ Terminator Ready Reaction Kit
Product Code. 16345272
Change view
Click to view available options
No. of Reactions:
100
Unit Size:
Each
Product Code. No. of Reactions Unit Size
16345272 100 Each
1 options
Product Code. 16345272 Supplier Applied Biosystems™ Supplier No. 4307176
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for working with GT- and G-rich and other difficult-to-sequence templates
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Description dGTP BigDye™ Terminator Ready Reaction Kit
No. of Reactions 100
Product Type dGTP BigDye™ Terminator Ready Reaction Kit