missing translation for 'onlineSavingsMsg'
Learn More

DHBS, MedChemExpress

Product Code. 16471569
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471569 Supplier Medchem Express Supplier No. HY15914
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DHBS is Used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of peroxidase in coupled enzymatic reactions.

DHBS is Used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of peroxidase in coupled enzymatic reactions.

Specifications

Chemical Name or Material DHBS
CAS 54970-72-8
Purity Grade Notes Research
Molecular Formula C6H3Cl2NaO4S
Synonym DCHBS
Solubility Information H2O : 100 mg/mL (377.29 mM; Need ultrasonic)
SMILES O=S(C1=CC(Cl)=CC(Cl)=C1O)(O[Na])=O
Molecular Weight (g/mol) 265.05
Formula Weight 265.05
Percent Purity 97.0%
Grade Research
Color White
Quantity 1 g
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.