missing translation for 'onlineSavingsMsg'
Learn More

Di-2-ethylhexylamine, 99%

Product Code. 10277640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10277640 1 L 1 L
10043360 250 mL 250mL
10125353 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10277640 Supplier Thermo Scientific Acros Supplier No. 271960010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-20-7 | C16H35N | 241.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-20-7
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
MDL Number MFCD00009489
InChI Key SAIKULLUBZKPDA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619
PubChem CID 7791
IUPAC Name 2-ethyl-N-(2-ethylhexyl)hexan-1-amine
SMILES CCCCC(CC)CNCC(CC)CCCC

Specifications

Melting Point -60.0°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 281.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3(CH2)3CH(C2H5)CH2)2NH
Refractive Index 1.4415 to 1.4435
Quantity 1 L
Beilstein 04,388
Specific Gravity 0.8
Solubility Information Solubility in water: <0.2g/L (20 c) in water. Other solubilities: soluble in hydrocarbons
Viscosity 3.7 mPa.s (20°C)
Formula Weight 241.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Di-2-ethylhexylamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Di-2-ethylhexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.