missing translation for 'onlineSavingsMsg'
Learn More

Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%

Product Code. 10467402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging Dimensione della confezione
10467402 1 g Glass bottle 1g
10320304 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10467402 Fornitore Thermo Scientific Acros N. del fornitore 363290010
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 52462-29-0 | C20H28Cl4Ru2 | 612.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 52462-29-0
Molecular Formula C20H28Cl4Ru2
Molecular Weight (g/mol) 612.39
MDL Number MFCD00064793
InChI Key LAXRNWSASWOFOT-UHFFFAOYSA-J
Synonym dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer
PubChem CID 10908223
IUPAC Name dichlororuthenium;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl

Specifica

Melting Point 250°C
Color Brown-Red to Orange-Red
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 612.39
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Di-μ-chlorobis(p-cymene)chlororuthenium(II)
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Di-mu-chlorobis(p-cymene)chlororuthenium(II)
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Indicazioni di pericolo
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Consigli di prudenza
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.