missing translation for 'onlineSavingsMsg'
Learn More

Di-Iso-Propyl Ether, Certified AR for Analysis, Stabilised with BHT, Fisher Chemical™

Product Code. 10657022 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Unit Size:
1 L
2.5 L
25 L
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10657022 1 L Amber glass bottle 1 L
10366180 2.5 L Amber glass bottle 2.5 L
10699103 5 L Metal can 5 L
10082923 25 L Metal can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10657022 Supplier Fisher Chemical Supplier No. D/5002/15
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

C6H14O, CAS Number-108-20-3, 2,2'-oxydipropane, 2-isopropoxypropane, bis isopropyl ether, diisopropylether, diisopropyl ether, diisopropyl oxide, ether, isopropyl, isopropylether, isopropyl ether, propane, 2,2'-oxybis, 2.5L, 68 deg.C, Colorless, 102.18g/mol, ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number 8880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Melting Point -85.5°C
Color Colorless
Boiling Point 68°C
Packaging Amber glass bottle
Quantity 1 L
Vapor Density 1.42
Formula Weight 102.18g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Isopropyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Supplemental information
  • MIXTURE LIST-Contain: Isopropyl ether, 2,6-Di-tert-butyl-p-cresol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.