missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl ether, 99+%, Extra Dry, AcroSeal™

Product Code. 10607764 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10607764 1 L 1 L
10573324 100 mL 100mL
11934701 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10607764 Supplier Thermo Scientific Acros Supplier No. 396910010
  • ONLINE EXCLUSIVE PRICE
    209.00 €
    191.00 € / 1 L
    Save 18.00 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-96-1 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-96-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00009461
InChI Key DURPTKYDGMDSBL-UHFFFAOYSA-N
Synonym di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
PubChem CID 8909
IUPAC Name 1-butoxybutane
SMILES CCCCOCCCC

Specifications

Melting Point -95.0°C
Color Colorless
Density 0.7680g/mL
Boiling Point 141.0°C
Flash Point 25°C
Assay Percent Range 99% min. (GC)
Packaging AcroSeal glass Bottle
Linear Formula [CH3(CH2)3]2O
Refractive Index 1.3978 to 1.3998
Quantity 1 L
Residue after Evaporation 0.0005% max.
Merck Index 15,1572
Specific Gravity 0.768
Solubility Information Solubility in water: 0.03% (20°C). Other solubilities: miscible with ethanol,ether,acetone
Viscosity 0.69 mPa.s (20°C)
Formula Weight 130.23
Percent Purity 99+%
Grade Extra Dry
Physical Form Liquid
Chemical Name or Material Di-n-butyl ether, Extra Dry
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Di-n-butyl ether
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.