missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl sulfate, 95%

Product Code. 11383376
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383376 10 g 10g
11393376 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383376 Supplier Thermo Scientific Alfa Aesar Supplier No. L12813.09
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 625-22-9 | C8H18O4S | 210.288 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-22-9
Molecular Formula C8H18O4S
Molecular Weight (g/mol) 210.288
MDL Number MFCD00059420
InChI Key LMEDOLJKVASKTP-UHFFFAOYSA-N
Synonym dibutylsulfate, di-n-butylsulfat, sulfuric acid, dibutyl ester, di-n-butylsulfate, di-n-butyl sulfate, dibutyl sulphate, di-n-butylsulfat german, sulfuric acid dibutyl ester, sulphuric acid dibutyl ester, dibutylsulfat
PubChem CID 12239
IUPAC Name dibutyl sulfate
SMILES CCCCOS(=O)(=O)OCCCC

Specifications

Density 1.06
Boiling Point 115°C to 116°C (6 mmHg)
Linear Formula (CH3CH2CH2CH2O)2SO2
Refractive Index 1.4205
Quantity 10 g
Beilstein 1707498
Formula Weight 210.29
Percent Purity 95%
Chemical Name or Material Di-n-butyl sulfate
compliance-icons
Product Identifier
  • Di-n-butyl sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Sulfuric acid, dibutyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.