missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl sulfide, 98%

Product Code. 11301977
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301977 100 mL 100mL
11311977 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301977 Supplier Thermo Scientific Alfa Aesar Supplier No. L07599.AE
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 544-40-1 | C8H18S | 146.29 g/mol

Di-n-butyl sulfide is generally used as an agricultural intermediate, a sulfiding agent, a refinery catalyst, a lubricant additive, a gas odorant and a processing aid in mining applications. It finds application as an internal standard in X-ray fluorescence spectrometry (XRF) measurements of sulfur in oils and other liquid hydrocarbon matrices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-butyl sulfide is generally used as an agricultural intermediate, a sulfiding agent, a refinery catalyst, a lubricant additive, a gas odorant and a processing aid in mining applications. It finds application as an internal standard in X-ray fluorescence spectrometry (XRF) measurements of sulfur in oils and other liquid hydrocarbon matrices.

Solubility
Silghtly miscible with water. Miscible with olive oil and almond oil.

Notes
Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-40-1
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00009468
InChI Key HTIRHQRTDBPHNZ-UHFFFAOYSA-N
Synonym dibutyl sulfide, butyl sulfide, n-butyl sulfide, butylthiobutane, di-n-butyl sulfide, butane, 1,1'-thiobis, butyl monosulfide, dibutyl thioether, thiononane-5, n-dibutyl sulfide
PubChem CID 11002
IUPAC Name 1-butylsulfanylbutane
SMILES CCCCSCCCC

Specifications

Melting Point -80°C
Density 0.839
Boiling Point 188°C to 189°C
Flash Point 76°C (168°F)
Odor Stench
Linear Formula CH3(CH2)3S(CH2)3CH3
Refractive Index 1.453
Quantity 100 mL
UN Number UN3334
Beilstein 1732829
Merck Index 14,1590
Solubility Information Silghtly miscible with water. Miscible with olive oil and almond oil.
Formula Weight 146.3
Percent Purity 98%
Chemical Name or Material Di-n-butyl sulfide
Show More Show Less
compliance-icons
Product Identifier
  • Di-n-butyl sulfide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Di-n-butyl sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.