missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylamine, 98+%

Product Code. 11479153
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479153 2500 mL 2500mL
11469153 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479153 Supplier Thermo Scientific Alfa Aesar Supplier No. A11671.0F
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-92-2 | C8H19N | 129.247 g/mol

Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.

Solubility
Miscible with water, ether, ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, carbon dioxide, zinc, iron and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-92-2
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

Melting Point -62°C
Density 0.767
Boiling Point 159°C to 160°C
Flash Point 39°C (102°F)
Assay Percent Range ≥98%
Odor Amine-like
Linear Formula [CH3(CH2)3]2NH
Refractive Index 1.4169
Quantity 2500 mL
UN Number UN2248
Beilstein 506001
Merck Index 14,3032
Solubility Information Miscible with water,ether,ethanol and acetone.
Formula Weight 129.25
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Di-n-butylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-n-butylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Di-n-butylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.