missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylamine, 99%

Product Code. 10627642 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
50 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
50mL
5mL
This item is not returnable. View return policy

Product Code. 10627642

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL | CAS: 111-92-2 | C8H19N | 129.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-92-2
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

Melting Point -62.0°C
Density 0.7600g/mL
Boiling Point 159.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H19N
Linear Formula [CH3(CH2)3]2NH
Refractive Index 1.4160 to 1.4180
Quantity 2.5 L
Beilstein 04, 157
Fieser 01,209
Merck Index 15, 3037
Specific Gravity 0.76
Solubility Information Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons
Molecular Weight (g/mol) 129.24
Viscosity 0.9 mPa.s (20°C)
Formula Weight 129.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Di-n-butylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-n-butylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.