missing translation for 'onlineSavingsMsg'
Learn More

Di-N-Butylamine Phosphate, for HPLC, Certified, Solution 1.0M, Fisher Chemical

Product Code. 10283180 Shop All Fisher Chemical Products
Click to view available options
:
100mL, Amber glass bottle
This item is not returnable. View return policy

Product Code. 10283180

Brand: Fisher Chemical D/1341/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 34655-69-1
Color Colorless
Packaging Amber glass bottle
Molecular Formula C8H22NO4P
MDL Number 42059
Quantity 100 mL
InChI Key GIXHHVDNJZSLBZ-UHFFFAOYSA-N
Molecular Weight (g/mol) 227.24
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.