missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl disulfide, tech. 85%

Product Code. 11435147
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435147 25 g 25g
11425147 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435147 Supplier Thermo Scientific Alfa Aesar Supplier No. B22568.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10496-15-8 | C12H26S2 | 234.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10496-15-8
Molecular Formula C12H26S2
Molecular Weight (g/mol) 234.46
MDL Number MFCD00027300
InChI Key GJPDBURPGLWRPW-UHFFFAOYSA-N
Synonym hexyl disulfide, dihexyl disulfide, di-n-hexyl disulfide, disulfide, dihexyl, dihexyl disulphide, 1-hexyldisulfanyl hexane, disulfide,dihexyl, dihexyl persulfide, n-hexyl disulfide, di-n-hexyldisulfide
PubChem CID 82675
IUPAC Name 1-(hexyldisulfanyl)hexane
SMILES CCCCCCSSCCCCCC

Specifications

Density 0.919
Flash Point >110°C (230°F)
Odor Unpleasant
Quantity 25 g
Formula Weight 234.47
Percent Purity 85%
Grade Technical
Chemical Name or Material Di-n-hexyl disulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.