missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl sulfide, 97%

Product Code. 11419096
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11419096

Brand: Thermo Scientific Alfa Aesar A17955.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2690-08-6 | C16H34S | 258.51 g/mol

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in water, 0.004678 mg/L @ 25°C (est)

Notes
Air & Moisture Sensitive. Store away from acids, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2690-08-6
MDL Number MFCD00009566

Specifications

Melting Point 0°C to 1°C
Density 0.844
Boiling Point 179°C to 180°C (10 mmHg)
Flash Point 160°C (320°F)
Odor Stench
Molecular Formula C12H17NO
Refractive Index 1.4624
Quantity 500 g
Beilstein 1701624
Synonym benzamide, n,n-diethyl-4-methyl, p-deta, n,n-diethyl-p-toluamide, p-toluamide, n,n-diethyl, p-deet, unii-i8bwy8k43u, i8bwy8k43u, n,n-diethyl-4-methyl-benzamide, p-toluamide,n-diethyl, n,n-diethyl-4-toluamide
InChI Key PUZORFQMRDHKBT-UHFFFAOYSA-N
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)C
IUPAC Name N,N-diethyl-4-methylbenzamide
Molecular Weight (g/mol) 191.27
PubChem CID 75946
Formula Weight 258.51
Percent Purity 97%
Chemical Name or Material Di-n-octyl sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.