missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl sulfide, 97%

Product Code. 11499086
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499086 25 g 25g
11409096 100 g 100g
11419096 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499086 Supplier Thermo Scientific Alfa Aesar Supplier No. A17955.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2690-08-6 | C16H34S | 258.51 g/mol

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in water, 0.004678 mg/L @ 25°C (est)

Notes
Air & Moisture Sensitive. Store away from acids, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2690-08-6
MDL Number MFCD00009566

Specifications

Melting Point 0°C to 1°C
Density 0.844
Boiling Point 179°C to 180°C (10 mmHg)
Flash Point 160°C (320°F)
Odor Stench
Molecular Formula C16H34S
Refractive Index 1.4624
Quantity 25 g
Beilstein 1701624
Synonym dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane
InChI Key LOXRGHGHQYWXJK-UHFFFAOYSA-N
SMILES CCCCCCCCSCCCCCCCC
IUPAC Name 1-octylsulfanyloctane
Molecular Weight (g/mol) 258.51
PubChem CID 75901
Formula Weight 258.51
Percent Purity 97%
Chemical Name or Material Di-n-octyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.