missing translation for 'onlineSavingsMsg'
Learn More

Di-n-pentyl sulfide, 97%

Product Code. 11361647
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11361647 25 g 25g
11351647 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361647 Supplier Thermo Scientific Alfa Aesar Supplier No. L04649.14
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 872-10-6 | C10H22S | 174.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-10-6
Molecular Formula C10H22S
Molecular Weight (g/mol) 174.35
MDL Number MFCD00027275
InChI Key JOZDADPMWLVEJK-UHFFFAOYSA-N
Synonym pentyl sulfide, diamyl sulfide, pentane, 1,1'-thiobis, amyl sulfide, n-amyl sulfide, dipentyl sulphide, di-n-pentyl sulfide, dipentyl sulfide, unii-61xjn3644r, di-n-pentyl sulphide
PubChem CID 13382
IUPAC Name 1-pentylsulfanylpentane
SMILES CCCCCSCCCCC

Specifications

Melting Point -51°C
Density 0.84
Boiling Point 228°C to 230°C
Flash Point 85°C (185°F)
Odor Stench
Linear Formula CH3(CH2)4S(CH2)4CH3
Refractive Index 1.456
Quantity 25 g
Beilstein 1698031
Formula Weight 174.35
Percent Purity 97%
Chemical Name or Material Di-n-pentyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.