missing translation for 'onlineSavingsMsg'
Learn More

Di-Norbudesonide(Mixture of Diastereomers), TRC

Product Code. 30137991
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137991 500 mg 500mg
30008501 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137991 Supplier Toronto Research Chemicals Supplier No. TRCH589650500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1040085-98-0
Molecular Formula C23 H30 O6
Molecular Weight (g/mol) 402.48
Synonym Pregna-1,4-diene-3,20-dione, 16,17-[ethylidenebis(oxy)]-11,21-dihydroxy-, (11β,16α)-, (11β,16α)-16,17-[Ethylidenebis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione, 16α,17-[(1RS)-Ethylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione, Budesonide acetaldehyde acetal, Budesonide Imp. B (EP)
IUPAC Name 16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione
SMILES CC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C23H30O6/c1-12-28-19-9-16-15-5-4-13-8-14(25)6-7-21(13,2)20(15)17(26)10-22(16,3)23(19,29-12)18(27)11-24/h6-8,12,15-17,19-20,24,26H,4-5,9-11H2,1-3H3/t12?,15-,16-,17-,19+,20+,21-,22-,23+/m0/s1
Formula Weight 402.2042
Percent Purity >95%
Chemical Name or Material 16 alpha,17-[(1RS)-Ethylidenebis(oxy)]-11 beta,21-dihydroxypregna-1,4-diene-3,20-dione
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.