missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-O-acetyl-L-tartaric Anhydride, TRC
Click to view available options
Quantity:
2 g
Unit Size:
2g
Specifications
Specifications
| Chemical Name or Material | Di-O-acetyl-L-tartaric Anhydride |
| CAS | 6283-74-5 |
| Molecular Formula | C8 H8 O7 |
| Quantity | 2 g |
| Synonym | 2,5-Furandione, 3,4-bis(acetyloxy)dihydro-, (3R,4R)-, 2,5-Furandione, 3,4-bis(acetyloxy)dihydro-, (3R-trans)-, Tartaric anhydride, diacetate (8CI), (3R,4R)-3,4-Bis(acetyloxy)dihydro-2,5-furandione, (+)-Diacetyl-L-tartaric anhydride, (2R,3R)-Di-O-acetyltartaric acid anhydride, (R,R)-2,3-Diacetoxy-1,4-butanedioic anhydride, (R,R)-O,O-Diacetyltartaric acid anhydride, (R,R)-O,O-Diacetyltartaric anhydride, Diacetyl-L-tartaric anhydride, Diacetyltartaric acid anhydride, Diacetyltartaric anhydride, L-Diacetyltartaric anhydride, NSC 7712, O,O'-Diacetyl-L-tartaric anhydride, Di-O-acetyl-L-tartaric Anhydride |
| InChI Formula | InChI=1S/C8H8O7/c1-3(9)13-5-6(14-4(2)10)8(12)15-7(5)11/h5-6H,1-2H3/t5-,6-/m1/s1 |
| SMILES | CC(=O)O[C@@H]1[C@@H](OC(=O)C)C(=O)OC1=O |
| IUPAC Name | [(3R,4R)-4-acetyloxy-2,5-dioxooxolan-3-yl] acetate |
| Molecular Weight (g/mol) | 216.14 |
| Formula Weight | 216.027 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction