missing translation for 'onlineSavingsMsg'
Learn More

Di-O-benzoyl L-Tartaric Acid, TRC

Product Code. 30367454
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367454 50 g 50g
30366769 5 g 5g
30358120 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367454 Supplier Toronto Research Chemicals Supplier No. TRCD41732650G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2743-38-6
Molecular Formula C18 H14 O8
Molecular Weight (g/mol) 358.3
Synonym (2R,3R)-2,3-Bis(benzoyloxy)butanedioic Acid, Ramipril Imp. M (EP), Dibenzoyltartric Acid, Butanedioic acid, 2,3-bis(benzoyloxy)-, (2R,3R)-, Butanedioic acid, 2,3-bis(benzoyloxy)-, [R-(R*,R*)]-, Tartaric acid, dibenzoate (6CI), Tartaric acid, dibenzoate, (-)- (8CI), (-)-2,3-Dibenzoyl-L-tartaric acid, (-)-Dibenzoyl-L-tartaric acid, (-)-Dibenzoyltartaric acid, (-)-L-Dibenzoyltartaric acid, (-)-O,O'-Dibenzoyl-L-tartaric acid, (-)-O,O'-Dibenzoyltartaric acid, (2R,3R)-2,3-Bis(benzoyloxy)succinic acid, (2R,3R)-Di-O-benzoyltartaric acid, (2R,3R)-O,O'-Dibenzoyltartaric acid, (R,R)-2,3-Dibenzoyloxysuccinic acid, (R,R)-O,O'-Dibenzoyltartaric acid, 2,3-Di-O-benzoyl-L-tartaric acid, 2,3-Dibenzoyl-L-tartaric acid, Dibenzoyl-(+)-tartaric acid, Dibenzoyl-L-tartaric acid, L-(-)-Dibenzoyltartaric acid, L-Dibenzoyltartaric acid, NSC 118224, NSC 338494, O,O-Dibenzoyl-(R,R)-tartaric acid, O,O-Dibenzoyltartaric acid, O,O'-Dibenzoyl-(2R,3R)-tartaric acid, O,O'-Dibenzoyl-(R,R)-tartaric acid, O2,O3-Dibenzoyl-L-tartaric acid
IUPAC Name (2R,3R)-2,3-dibenzoyloxybutanedioic acid
SMILES OC(=O)[C@H](OC(=O)c1ccccc1)[C@@H](OC(=O)c2ccccc2)C(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C18H14O8/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12/h1-10,13-14H,(H,19,20)(H,21,22)/t13-,14-/m1/s1
Formula Weight 358.0689
Chemical Name or Material (2R,3R)-2,3-Bis(benzoyloxy)butanedioic Acid (Dibenzoyltartric Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.