missing translation for 'onlineSavingsMsg'
Learn More

Di-p-toluoyl-D-tartaric acid, 98%

Product Code. 11337976
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337976 100 g 100g
11317976 5 g 5g
11327976 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337976 Supplier Thermo Scientific Alfa Aesar Supplier No. L13223.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32634-68-7 | C20H18O8 | 386.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32634-68-7
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00008552
InChI Key CMIBUZBMZCBCAT-HOTGVXAUSA-N
Synonym di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid
PubChem CID 263211
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 169°C to 171°C
Quantity 100 g
Optical Rotation +139° (c=1 in Ethanol)
Beilstein 3225584
IUPAC Name (2S,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid
Formula Weight 386.36
Percent Purity 98%
Chemical Name or Material Di-p-toluoyl-D-tartaric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.