missing translation for 'onlineSavingsMsg'
Learn More

Di-p-toluoyl-L-tartaric acid, 98%

Product Code. 11436126
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436126 25 g 25g
11426126 5 g 5g
11446126 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436126 Supplier Thermo Scientific Alfa Aesar Supplier No. B22972.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32634-66-5 | C20H18O8 | 386.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32634-66-5
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00064440
InChI Key CMIBUZBMZCBCAT-IYBDPMFKSA-N
Synonym 2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r
PubChem CID 12297760
IUPAC Name (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 166°C to 171°C
Beilstein 2022481
Formula Weight 386.36
Percent Purity 98%
Quantity 25 g
Optical Rotation −139° (c=1 in Ethanol)
Chemical Name or Material Di-p-toluoyl-L-tartaric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.